Merck
  • Mogrol represents a novel leukemia therapeutic, via ERK and STAT3 inhibition.
Mogrol represents a novel leukemia therapeutic, via ERK and STAT3 inhibition.
American journal of cancer research (2015-06-24)
Can Liu, Yan Zeng, Long-Hai Dai, Tian-Yu Cai, Yue-Ming Zhu, De-Quan Dou, Lan-Qing Ma, Yuan-Xia Sun
ABSTRACT
Unlike solid tumors, the primary strategy for leukemia treatment is chemotherapy. However, leukemia chemotherapy is associated with adverse drug effects and drug resistance. Therefore, it is imperative to identify novel agents that effectively treat leukemia while minimizing adverse effects. The Raf/MEK/extracellular regulated kinase (ERK) and signal transducer and activator of transcription 3 (STAT3) pathways have been implicated in leukemia carcinogenesis, and provide novel molecular targets for therapeutic intervention in cancer. Mogrol, a biometabolite of mogrosides found in Siraitia grosvenorii, has exhibited anti-cancer activities; however, the underlying mechanism of this effect remains unclear. To clarify its anti-cancer activity and mechanism of action, we treated K562 leukemia cells with mogrol. Mogrol suppressed leukemia cell growth via inhibition of the ERK1/2 and STAT3 pathways, in particular, through the suppression of p-ERK1/2 and p-STAT3. Inhibition of these pathways suppressed Bcl-2 expression, thereby inducing K562 cell apoptosis. Furthermore, mogrol enhanced p21 expression, resulting in G0/G1 cell cycle arrest. The findings provide new perspectives regarding the role of mogrol in leukemia treatment.
Sigma-Aldrich
Mogrol, ≥98% (HPLC)
Pricing and availability is not currently available.