Merck
The use of misoprostol in obstetrics and gynaecology.
BJOG : an international journal of obstetrics and gynaecology (2009-09-26)
A Elati, A D Weeks
ABSTRACT
Misoprostol, although originally introduced as a therapy for gastric ulcers, is now widely used in reproductive health. For some indications it is now the optimal choice, whilst for others it provides an important alternative, especially in low-resource settings. The optimal dose varies widely from 20 to 600 mcg depending on the indication and gestation. Use of the correct dose is important, too low a dose will be ineffective and overdosage can be dangerous for mother and baby. Evidence-based information about the safest regimens for multiple pregnancy indications are therefore provided in this review.
Sigma-Aldrich
Misoprostol free acid, ≥90%