Merck
  • RNA interference-based (RNAi) suppression of AtMPK6, an Arabidopsis mitogen-activated protein kinase, results in hypersensitivity to ozone and misregulation of AtMPK3.
RNA interference-based (RNAi) suppression of AtMPK6, an Arabidopsis mitogen-activated protein kinase, results in hypersensitivity to ozone and misregulation of AtMPK3.
Environmental pollution (Barking, Essex : 1987) (2005-06-21)
Godfrey P Miles, Marcus A Samuel, Yuelin Zhang, Brian E Ellis
ABSTRACT
The recent increase in tropospheric ozone (O(3)) concentrations promotes additional oxidative stress through the production of reactive oxygen species (ROS) in plant tissues, resulting in the activation of genes whose products enable the stressed cells to retain their integrity and function. This response is made possible by an integration of highly regulated signaling networks that mediate the perception of, and response to, this oxidative assault. In Arabidopsis thaliana, ROS-induced signaling has been shown to flow through a protein phosphorylation cascade involving the mitogen-activated protein kinases (MAPKs) AtMPK3 (MPK3) and AtMPK6 (MPK6). We found that RNAi-mediated silencing of MPK6 renders the plant more sensitive to ozone, as determined by visible leaf damage. The MPK6-RNAi genotype also displayed a more intense and prolonged activation of MPK3 compared to that of WT plants. An MPK3 loss-of-function genotype is similarly very sensitive to ozone, and displays an abnormally prolonged MPK6 activation profile, suggesting reciprocity in regulation between these two MAPKs.