Merck
  • Discrete proteolysis of neuronal calcium sensor-1 (NCS-1) by mu-calpain disrupts calcium binding.
Discrete proteolysis of neuronal calcium sensor-1 (NCS-1) by mu-calpain disrupts calcium binding.
Cell calcium (2009-09-08)
Courtney Blachford, Andjelka Celić, Edward T Petri, Barbara E Ehrlich
ABSTRACT
Neuronal calcium sensor-1 (NCS-1) is a high-affinity, low-capacity Ca(2+)-binding protein expressed in many cell types. We previously showed that NCS-1 interacts with inositol 1,4,5-trisphosphate receptor (InsP(3)R) and modulates Ca(2+)-signaling by enhancing InsP3-dependent InsP(3)R channel activity and intracellular Ca(2+) transients. Recently we reported that the chemotherapeutic agent, paclitaxel (taxol) triggers mu-calpain dependent proteolysis of NCS-1, leading to reduced Ca(2+)-signaling within the cell. Degradation of NCS-1 may be critical in the induction of peripheral neuropathy associated with taxol treatment for breast and ovarian cancer. To begin to design strategies to protect NCS-1, we treated NCS-1 with mu-calpain in vitro and identified the cleavage site by N-terminal sequencing and MALDI mass spectroscopy. mu-Calpain cleavage of NCS-1 occurs within an N-terminal pseudoEF-hand domain, which by sequence analysis appears to be unable to bind Ca(2+). Our results suggest a role for this pseudoEF-hand in stabilizing the three functional EF-hands within NCS-1. Using isothermal titration calorimetry (ITC) we found that loss of the pseudoEF-hand markedly decreased NCS-1's affinity for Ca(2+). Physiologically, this significant decrease in Ca(2+) affinity may render NCS-1 incapable of responding to changes in Ca(2+) levels in vivo. The reduced ability of mu-calpain treated NCS-1 to bind Ca(2+) may explain the altered Ca(2+) signaling in the presence of taxol and suggests a strategy for therapeutic intervention of peripheral neuropathy in cancer patients undergoing taxol treatment.