Merck
  • The reliability of the isotropic fractionator method for counting total cells and neurons.
The reliability of the isotropic fractionator method for counting total cells and neurons.
Journal of neuroscience methods (2019-08-09)
Kleber Neves, Daniel Menezes Guimarães, Danielle Rayêe, Bruna Valério-Gomes, Pamela Meneses Iack, Roberto Lent, Bruno Mota
ABSTRACT
The Isotropic Fractionator (IF) is a method to determine the cellular composition of nervous tissue. It has been mostly applied to assess variation across species, where differences are expected to be large enough not to be masked by methodological error. However, understanding the sources of variation in the method is important if the goal is to detect smaller differences, for example, in same-species comparisons. Comparisons between different mice strains suggest that the IF is consistent enough to detect these differences. Nevertheless, the reliability of the method has not yet been examined directly. In this study, we evaluate the reliability of the method for the determination of cellular and neuronal numbers of Swiss mice. We performed repeated cell counts of the same material by different experimenters to quantify different sources of variation. In total cell counts, we observed that for the cerebral cortex most of the variance was at the counter level. For the cerebellum, most of the variance is attributed to the sample itself. As for neurons, random error along with the immunostaining correspond to most of the variation, both in the cerebral cortex and in the cerebellum. Test-retest reliability coefficients were relatively high, especially for cell counts. Although biases between counters and random variation in staining could be problematic when aggregating data from different sources, we offer practical suggestions to improve the reliability of the method. While small, this study is a most needed step towards more precise measurement of the brain's cellular composition.