Merck
  • Kdm4c is Recruited to Mitotic Chromosomes and Is Relevant for Chromosomal Stability, Cell Migration and Invasion of Triple Negative Breast Cancer Cells.
Kdm4c is Recruited to Mitotic Chromosomes and Is Relevant for Chromosomal Stability, Cell Migration and Invasion of Triple Negative Breast Cancer Cells.
Breast cancer : basic and clinical research (2018-08-08)
Jeison Garcia, Fernando Lizcano
ABSTRACT
Members of the jumonji-containing lysine demethylase protein family have been associated with cancer development, although their specific roles in the evolution of tumor cells remain unknown. This work examines the effects of lysine demethylase 4C (KDM4C) knockdown on the behavior of a triple-negative breast cancer cell line. KDM4C expression was knocked-down by siRNA and analyzed by Western blot and immunofluorescence. HCC38 cell proliferation was examined by MTT assay, while breast cancer cells' migration and invasion were tested in Transwell format by chemotaxis. Immunofluorescence assays showed that KDM4C, which is a key protein for modulating histone demethylation and chromosome stability through the distribution of genetic information, is located at the chromosomes during mitosis. Proliferation assays demonstrated that KDM4C is important for cell survival, while Transwell migration and invasion assays indicated that this protein is relevant for cancer progression. These data indicate that KDM4C is relevant for breast cancer progression and highlight its importance as a potential therapeutic target.