Merck
  • Arthritis flares mediated by tissue-resident memory T cells in the joint.
Arthritis flares mediated by tissue-resident memory T cells in the joint.
Cell reports (2021-10-28)
Margaret H Chang, Anaïs Levescot, Nathan Nelson-Maney, Rachel B Blaustein, Kellen D Winden, Allyn Morris, Alexandra Wactor, Spoorthi Balu, Ricardo Grieshaber-Bouyer, Kevin Wei, Lauren A Henderson, Yoichiro Iwakura, Rachael A Clark, Deepak A Rao, Robert C Fuhlbrigge, Peter A Nigrovic
ABSTRACT
Rheumatoid arthritis is a systemic autoimmune disease, but disease flares typically affect only a subset of joints, distributed in a distinctive pattern for each patient. Pursuing this intriguing pattern, we show that arthritis recurrence is mediated by long-lived synovial resident memory T cells (TRM). In three murine models, CD8+ cells bearing TRM markers remain in previously inflamed joints during remission. These cells are bona fide TRM, exhibiting a failure to migrate between joints, preferential uptake of fatty acids, and long-term residency. Disease flares result from TRM activation by antigen, leading to CCL5-mediated recruitment of circulating effector cells. Correspondingly, TRM depletion ameliorates recurrence in a site-specific manner. Human rheumatoid arthritis joint tissues contain a comparable CD8+-predominant TRM population, which is most evident in late-stage leukocyte-poor synovium, exhibiting limited T cell receptor diversity and a pro-inflammatory transcriptomic signature. Together, these findings establish synovial TRM as a targetable mediator of disease chronicity in autoimmune arthritis.
Sigma-Aldrich
Diphtheria Toxin from Corynebacterium diphtheriae, lyophilized powder, Product is in unnicked form
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Please contact Customer Service for Availability
CA$108.00
1 L
Please contact Customer Service for Availability
CA$586.00
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Please contact Customer Service for Availability
CA$108.00
1 L
Please contact Customer Service for Availability
CA$586.00
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Please contact Customer Service for Availability
CA$108.00
1 L
Please contact Customer Service for Availability
CA$586.00
Sigma-Aldrich
Anti-CD3, T Cell antibody produced in rabbit, whole antiserum
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Please contact Customer Service for Availability
CA$108.00
1 L
Please contact Customer Service for Availability
CA$586.00
Sigma-Aldrich
Albumin methylated from bovine serum
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Please contact Customer Service for Availability
CA$108.00
1 L
Please contact Customer Service for Availability
CA$586.00
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Please contact Customer Service for Availability
CA$108.00
1 L
Please contact Customer Service for Availability
CA$586.00
Sigma-Aldrich
Anti-CD69 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Please contact Customer Service for Availability
CA$108.00
1 L
Please contact Customer Service for Availability
CA$586.00