Merck
  • Mechanical homeostasis of liver sinusoid is involved in the initiation and termination of liver regeneration.
Mechanical homeostasis of liver sinusoid is involved in the initiation and termination of liver regeneration.
Communications biology (2021-04-09)
Jun Ishikawa, Makoto Takeo, Ayako Iwadate, Junko Koya, Miho Kihira, Masamitsu Oshima, Yuki Suzuki, Kazushi Taniguchi, Ayaka Kobayashi, Takashi Tsuji
ABSTRACT
Organogenesis and regeneration are fundamental for developmental progress and are associated with morphogenesis, size control and functional properties for whole-body homeostasis. The liver plays an essential role in maintaining homeostasis of the entire body through various functions, including metabolic functions, detoxification, and production of bile, via the three-dimensional spatial arrangement of hepatic lobules and has high regenerative capacity. The regeneration occurs as hypertrophy, which strictly controls the size and lobule structure. In this study, we established a three-dimensional sinusoidal network analysis method and determined valuable parameters after partial hepatectomy by comparison to the static phase of the liver. We found that mechanical homeostasis, which is crucial for organ morphogenesis and functions in various phenomena, plays essential roles in liver regeneration for both initiation and termination of liver regeneration, which is regulated by cytokine networks. Mechanical homeostasis plays critical roles in the initiation and termination of organogenesis, tissue repair and organ regeneration in coordination with cytokine networks.