Merck
  • Insulin signaling regulates longevity through protein phosphorylation in Caenorhabditis elegans.
Insulin signaling regulates longevity through protein phosphorylation in Caenorhabditis elegans.
Nature communications (2021-07-29)
Wen-Jun Li, Chen-Wei Wang, Li Tao, Yong-Hong Yan, Mei-Jun Zhang, Ze-Xian Liu, Yu-Xin Li, Han-Qing Zhao, Xue-Mei Li, Xian-Dong He, Yu Xue, Meng-Qiu Dong
ABSTRACT
Insulin/IGF-1 Signaling (IIS) is known to constrain longevity by inhibiting the transcription factor FOXO. How phosphorylation mediated by IIS kinases regulates lifespan beyond FOXO remains unclear. Here, we profile IIS-dependent phosphorylation changes in a large-scale quantitative phosphoproteomic analysis of wild-type and three IIS mutant Caenorhabditis elegans strains. We quantify more than 15,000 phosphosites and find that 476 of these are differentially phosphorylated in the long-lived daf-2/insulin receptor mutant. We develop a machine learning-based method to prioritize 25 potential lifespan-related phosphosites. We perform validations to show that AKT-1 pT492 inhibits DAF-16/FOXO and compensates the loss of daf-2 function, that EIF-2α pS49 potently inhibits protein synthesis and daf-2 longevity, and that reduced phosphorylation of multiple germline proteins apparently transmits reduced DAF-2 signaling to the soma. In addition, an analysis of kinases with enriched substrates detects that casein kinase 2 (CK2) subunits negatively regulate lifespan. Our study reveals detailed functional insights into longevity.
Product Description
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)