Merck
  • Gentiopicroside ameliorates ethanol-induced gastritis via regulating MMP-10 and pERK1/2 signaling.
Gentiopicroside ameliorates ethanol-induced gastritis via regulating MMP-10 and pERK1/2 signaling.
International immunopharmacology (2020-12-10)
Ying Chang, Yun Tian, Dan Zhou, Li Yang, Tie-Ming Liu, Zhen-Guo Liu, Si-Wang Wang
ABSTRACT
Excessive ethanol consumption results in gastric mucosa damage, which could further develop into chronic gastritis, peptic ulcer, and gastric cancer in humans. Gentiopicroside (GPS), a major active component of Gentianae Macrophyllae radix, was reported to play a critical role in anti-inflammation. In the study, we aimed to investigate the functional role and underlying mechanism of GPS in ethanol-induced gastritis. A model of gastritis was created by ethanol in C57BL/6 mice. Enzyme-linked immunosorbent assay was used to determine the concentration of TNF-α, IL-1β, IL-8, and IL-10. We found that GPS treatment significantly ameliorated ethanol-induced gastritis in mice, with lower production of pro-inflammatory cytokine TNF-α, IL-1β, and IL-8 and higher levels of anti-inflammatory cytokine IL-10. The anti-inflammatory effect of GPS was further confirmed in vitro in ethanol-treated human gastric mucosal GES cells. Mechanistically, we demonstrated that GPS regulated matrix metallopeptidase expression and pERK1/2 signaling. Knockdown of matrix metallopeptidase 10 (MMP-10) greatly improved cell survival and suppressed inflammatory response in ethanol-treated GES cells. Moreover, inhibition of pERK1/2 signaling using U0126 decreased the expression of MMP-10 in ethanol-induced gastritis. U0126 treatment also suppressed the expression of TNF-α, IL-1β, and IL-8, and enhanced IL-10 expression in mice gastric mucosa. Taken together, our findings suggest that GPS ameliorates ethanol-induced gastritis via regulating MMP-10 and pERK1/2 signaling, which might provide a promising therapeutic drug for ethanol-induced gastritis.
Sigma-Aldrich
MISSION® esiRNA, targeting human MMP10
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder