Merck
  • Molecular docking-aided identification of small molecule inhibitors targeting β-catenin-TCF4 interaction.
Molecular docking-aided identification of small molecule inhibitors targeting β-catenin-TCF4 interaction.
iScience (2021-06-19)
Joo-Leng Low, Weina Du, Tenzin Gocha, Gokce Oguz, Xiaoqian Zhang, Ming Wei Chen, Srdan Masirevic, Daniel Guo Rong Yim, Iain Bee Huat Tan, Adaikalavan Ramasamy, Hao Fan, Ramanuj DasGupta
ABSTRACT
Here we report a molecular docking-based approach to identify small molecules that can target the β-catenin (β-cat)-TCF4 protein-protein interaction (PPI), a key effector complex for nuclear Wnt signaling activity. Specifically, we developed and optimized a computational model of β-cat using publicly available β-cat protein crystal structures, and existing β-cat-TCF4 interaction inhibitors as the training set. Using our computational model to an in silico screen predicted 27 compounds as good binders to β-cat, of which 3 were identified to be effective against a Wnt-responsive luciferase reporter. In vitro functional validation experiments revealed GB1874 as an inhibitor of the Wnt pathway that targets the β-cat-TCF4 PPI. GB1874 also affected the proliferation and stemness of Wnt-addicted colorectal cancer (CRC) cells in vitro. Encouragingly, GB1874 inhibited the growth of CRC tumor xenografts in vivo, thus demonstrating its potential for further development into therapeutics against Wnt-associated cancer indications.
Sigma-Aldrich
Monoclonal Anti-β-Catenin antibody produced in mouse, clone 15B8, ascites fluid