Merck
  • Characterization of arylsulfatase formed by derepressed synthesis in Citrobacter braakii.
Characterization of arylsulfatase formed by derepressed synthesis in Citrobacter braakii.
Bioscience, biotechnology, and biochemistry (2006-06-24)
Toyokazu Miura, Kenji Okamoto, Hideshi Yanase
ABSTRACT
Arylsulfatase activity was detected in a bacterial strain, Citrobacter braakii 69-b, isolated from soil by enrichment cultivation using porcine gastric mucin. The production of arylsulfatase was derepressed markedly in a synthetic medium by the addition of tyramine. The purified enzyme hydrolyzed 4-nitrophenyl sulfate, 4-nitrocatechol sulfate, and 3-indoxyl sulfate, and was classified as type I arylsulfatase.
Sigma-Aldrich
Sulfatase from Aerobacter aerogenes, Type VI, buffered aqueous glycerol solution, 2-5 units/mg protein (biuret), 10-20 units/mL
Sigma-Aldrich
4-Nitrocatechol sulfate dipotassium salt, crystalline