Merck
  • Cyclooxygenase-2 inhibitor (SC-236) suppresses activator protein-1 through c-Jun NH2-terminal kinase.
Cyclooxygenase-2 inhibitor (SC-236) suppresses activator protein-1 through c-Jun NH2-terminal kinase.
Gastroenterology (2003-12-31)
Benjamin Chun-Yu Wong, Xiao Hua Jiang, Marie C m Lin, Shui Ping Tu, Jian Tao Cui, Shi Hu Jiang, Wai Man Wong, Man Fung Yuen, Shiu Kum Lam, Hsiang Fu Kung
ABSTRACT
Aspirin exerts antitumor effect partly through blocking tumor promoter-induced activator protein-1 (AP-1) activation. The aim of this study is to determine how specific COX-2 inhibitor SC-236 mediates antitumor effect by modulation of AP-1-signaling pathway. AP-1 transcriptional activity and DNA-binding activity were detected by luciferase reporter assay and gel shift assay, separately. Mitogen-activated protein kinase (MAPK) activation was determined by Western blot and in vitro kinase assay. Antisense oligonucleotide against c-Jun-N-terminal kinase (JNK) was used to suppress JNK expression. We showed that SC-236 inhibited 12-O-tetradecanoylphorbol-13-acetate (PMA)-induced cell transformation in a dose-dependent manner in JB6 cells. At a dose range (12.5-50 micromol/L) that inhibited cell transformation, SC-236 also inhibited anchorage-independent cell growth and AP-1-activation in 3 gastric cancer cells, independent of COX-prostaglandin synthesis. SC-236 down-regulated c-Jun-NH2-terminal kinase phosphorylation and activity. Suppression of JNK activity reversed the inhibitory effect on AP-1 activity by SC-236 and suppressed gastric cancer cell growth, indicating that the inhibitory effect of SC-236 on AP-1 activation and cell growth was through interaction with JNK. The inhibitory effect on JNK-c-Jun/AP-1 activation contributes to the antitumor effect of COX-2-specific inhibitor, and inhibition of JNK activation may have a therapeutic benefit against gastric cancer.
Sigma-Aldrich
SC-236, ≥98% (HPLC)