Merck
  • Differential expression of full-length and NH2 terminally truncated FAM134B isoforms in normal physiology and cancer.
Differential expression of full-length and NH2 terminally truncated FAM134B isoforms in normal physiology and cancer.
American journal of physiology. Gastrointestinal and liver physiology (2020-10-15)
Umur Keles, Evin Iscan, Huriye Erbak Yilmaz, Gökhan Karakülah, Aslı Suner, Erhan Bal, Nilgun Tasdemir, Ayse Derya Cavga, Umut Ekin, Zeynep Mutlu, Sila Kahyaoglu, Muhittin A Serdar, Nese Atabey, Mehmet Ozturk
ABSTRACT
Selective autophagy of the endoplasmic reticulum (ER), namely ER-phagy, is mediated by ER-localized receptors, which are recognized and sequestered by GABARAP/LC3B-decorated phagophores and transferred to lysosomes for degradation. Being one such receptor, FAM134B plays critical roles in cellular processes such as protein quality control and neuronal survival. FAM134B has also been associated with different cancers, although its exact role remains elusive. We report here that the FAM134B gene encodes not one but at least two different protein isoforms: the full-length and the NH2 terminally truncated forms. Their relative expression shows extreme variation, both within normal tissues and among cancer types. Expression of full-length FAM134B is restricted to the brain, testis, spleen, and prostate. In contrast, NH2 terminally truncated FAM134B is dominant in the heart, skeletal muscle, kidney, pancreas, and liver. We compared wild-type and knockout mice to study the role of the Fam134b gene in starvation. NH2 terminally truncated FAM134B-2 was induced in the liver, skeletal muscle, and heart but not in the pancreas and stomach following starvation. Upon starvation, Fam134b-/- mice differed from wild-type mice by less weight loss and less hyperaminoacidemic and hypocalcemic response but increased levels of serum albumin, total serum proteins, and α-amylase. Interestingly, either NH2 terminally truncated FAM134B or both isoforms were downregulated in liver, lung, and colon cancers. In contrast, upregulation was observed in stomach and chromophobe kidney cancers.NEW & NOTEWORTHY We reported tissues expressing FAM134B-2 such as the kidney, muscle, heart, and pancreas, some of which exhibit stimulated expression upon nutrient starvation. We also demonstrated the effect of Fam134b deletion during ad libitum and starvation conditions. Resistance to weight loss and hypocalcemia, accompanied by an increase in serum albumin and α-amylase levels, indicate critical roles of Fam134b in physiology. Furthermore, the differential expression of FAM134B isoforms was shown to be significantly dysregulated in human cancers.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-FAM134B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody