Merck
  • Macaques Exhibit Implicit Gaze Bias Anticipating Others' False-Belief-Driven Actions via Medial Prefrontal Cortex.
Macaques Exhibit Implicit Gaze Bias Anticipating Others' False-Belief-Driven Actions via Medial Prefrontal Cortex.
Cell reports (2020-04-03)
Taketsugu Hayashi, Ryota Akikawa, Keisuke Kawasaki, Jun Egawa, Takafumi Minamimoto, Kazuto Kobayashi, Shigeki Kato, Yukiko Hori, Yuji Nagai, Atsuhiko Iijima, Toshiyuki Someya, Isao Hasegawa
ABSTRACT
The ability to infer others' mental states is essential to social interactions. This ability, critically evaluated by testing whether one attributes false beliefs (FBs) to others, has been considered to be uniquely hominid and to accompany the activation of a distributed brain network. We challenge the taxon specificity of this ability and identify the causal brain locus by introducing an anticipatory-looking FB paradigm combined with chemogenetic neuronal manipulation in macaque monkeys. We find spontaneous gaze bias of macaques implicitly anticipating others' FB-driven actions. Silencing of the medial prefrontal neuronal activity with inhibitory designer receptor exclusively activated by designer drugs (DREADDs) specifically eliminates the implicit gaze bias while leaving the animals' visually guided and memory-guided tracking abilities intact. Thus, neuronal activity in the medial prefrontal cortex could have a causal role in FB-attribution-like behaviors in the primate lineage, emphasizing the importance of probing the neuronal mechanisms underlying theory of mind with relevant macaque animal models.