Merck
  • Methadone interrupts neural growth and function in human cortical organoids.
Methadone interrupts neural growth and function in human cortical organoids.
Stem cell research (2020-11-03)
Hang Yao, Wei Wu, Ines Cerf, Helen W Zhao, Juan Wang, Priscilla D Negraes, Alysson R Muotri, Gabriel G Haddad
ABSTRACT
Prenatal opioids exposure can lead to both neonatal abstinence syndrome in newborns and neurological deficits later in life. Although opioids have been well studied in general, the cellular and molecular mechanisms by which opioids affect human fetal brain development has not been well understood. In this work, we have taken advantage of a human 3D-brain cortical organoid (hCO) that facilitated enormously the investigation of early human brain development. Using imaging, immunofluorescence, multi-electrode array (MEA) and patch clamp recording techniques, we have investigated the effect of methadone, a frequently used opioid during pregnancy, on early neural development, including neuronal growth, neural network activity and synaptic transmission in hCOs. Our results demonstrated that methadone dose-dependently halted the growth of hCOs and induced organoid disintegration after a prolonged exposure. In addition, methadone dose-dependently suppressed the firing of spontaneous action potentials in hCOs and this suppression could be reversed upon methadone withdrawal in hCOs treated with lower dosages. Further investigation using patch clamp whole cell configuration revealed that, at clinically relevant concentrations, methadone decreased the frequency and amplitude of excitatory postsynaptic currents in neurons, indicating a critical role of methadone in weakening synaptic transmission in neural networks in hCOs. In addition, methadone significantly attenuated the voltage-dependent Na+ current in hCOs. We conclude that methadone interrupts neural growth and function in early brain development.
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
CTAP, ≥95%