- Phenyl sulfate, indoxyl sulfate and p-cresyl sulfate decrease glutathione level to render cells vulnerable to oxidative stress in renal tubular cells.
Phenyl sulfate, indoxyl sulfate and p-cresyl sulfate decrease glutathione level to render cells vulnerable to oxidative stress in renal tubular cells.
In chronic kidney disease patients, oxidative stress is generally associated with disease progression and pathogenesis of its comorbidities. Phenyl sulfate is a protein-bound uremic solute, which accumulates in chronic kidney disease patients, but little is known about its nature. Although many reports revealed that protein-bound uremic solutes induce reactive oxygen species production, the effects of these solutes on anti-oxidant level have not been well studied. Therefore, we examined the effects of protein-bound uremic solutes on glutathione levels. As a result, indoxyl sulfate, phenyl sulfate, and p-cresyl sulfate decreased glutathione levels in porcine renal tubular cells. Next we examined whether phenyl sulfate-treated cells becomes vulnerable to oxidative stress. In phenyl sulfate-treated cells, hydrogen peroxide induced higher rates of cell death than in control cells. Buthionine sulfoximine, which is known to decrease glutathione level, well mimicked the effect of phenyl sulfate. Finally, we evaluated a mixture of indoxyl sulfate, phenyl sulfate, and p-cresyl sulfate at concentrations comparable to the serum concentrations of hemodialysis patients, and we confirmed its decreasing effect on glutathione level. In conclusion, indoxyl sulfate, phenyl sulfate, and p-cresyl sulfate decrease glutathione levels, rendering the cells vulnerable to oxidative stress.