MilliporeSigma
  • Novel 199 base pair NEFH promoter drives expression in retinal ganglion cells.
Novel 199 base pair NEFH promoter drives expression in retinal ganglion cells.
Scientific reports (2020-10-07)
Sophia Millington-Ward, Naomi Chadderton, Megan Berkeley, Laura K Finnegan, Killian S Hanlon, Matthew Carrigan, Peter Humphries, Paul F Kenna, Arpad Palfi, G Jane Farrar
ABSTRACT
Retinal ganglion cells (RGCs) are known to be involved in several ocular disorders, including glaucoma and Leber hereditary optic neuropathy (LHON), and hence represent target cells for gene therapies directed towards these diseases. Restricting gene therapeutics to the target cell type in many situations may be preferable compared to ubiquitous transgene expression, stimulating researchers to identify RGC-specific promoters, particularly promoter sequences that may also be appropriate in size to fit readily into recombinant adeno associated viral (AAV) vectors, the vector of choice for many ocular gene therapies. In the current study we analysed EGFP expression driven by various sequences of the putative human NEFH promoter in order to define sequences required for preferential expression in RGCs. EGFP expression profiles from four different potential NEFH promoter constructs were compared in vivo in mice using retinal histology and mRNA expression analysis. Notably, two efficient promoter sequences, one comprising just 199 bp, are presented in the study.