Merck
  • Facile synthesis of magnetic sulfonated covalent organic framework composites for simultaneous dispersive solid-phase extraction and determination of β-agonists and fluoroquinolones in food samples.
Facile synthesis of magnetic sulfonated covalent organic framework composites for simultaneous dispersive solid-phase extraction and determination of β-agonists and fluoroquinolones in food samples.
Food chemistry (2020-11-07)
Kai Hu, Yanmei Shi, Weixia Zhu, Junlan Cai, Wenjie Zhao, Huahui Zeng, Zhenqiang Zhang, Shusheng Zhang
ABSTRACT
In this work, an efficient method for the determination of β-agonists and fluoroquinolones was established, based on a mixed-mode sorbent of magnetic sulfonated covalent organic framework composites. By coupling with HPLC-MS/MS, the main factors that affect the extraction procedure were optimized. Under the optimal conditions, the proposed HPLC-MS/MS method was successfully utilized for the extraction of β-agonists and fluoroquinolones in milk and pork meat samples. The method showed good linearities (R2 ≥ 0.9916), and low LOQs of 0.1-0.2 ng g-1 for β-agonists and fluoroquinolones. The adsorption mechanism was investigated with the assistance of quantum chemistry calculation method, and it is worth noting that the sorbent relied mainly on the multiple adsorption mechanisms, including π-π stacking, hydrophobic, electrostatic attraction and hydrogen-bonding interactions. This work not only provides a simple method for the preparation of a mixed-mode sorbent, but also a routine analysis strategy for monitoring the illegal use of β-agonists and fluoroquinolones.
Product Description
Supelco
Mabuterol hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Enrofloxacin, ≥99.0%