Merck
  • A role for E2F activities in determining the fate of Myc-induced lymphomagenesis.
A role for E2F activities in determining the fate of Myc-induced lymphomagenesis.
PLoS genetics (2009-09-15)
Rachel E Rempel, Seiichi Mori, Maura Gasparetto, Michele A Glozak, Eran R Andrechek, Steven B Adler, Nina M Laakso, Anand S Lagoo, Robert Storms, Clay Smith, Joseph R Nevins
ABSTRACT
The phenotypic heterogeneity that characterizes human cancers reflects the enormous genetic complexity of the oncogenic process. This complexity can also be seen in mouse models where it is frequently observed that in addition to the initiating genetic alteration, the resulting tumor harbors additional, somatically acquired mutations that affect the tumor phenotype. To investigate the role of genetic interactions in the development of tumors, we have made use of the Emu-myc model of pre-B and B cell lymphoma. Since various studies point to a functional interaction between Myc and the Rb/E2F pathway, we have investigated the role of E2F activities in the process of Myc-induced lymphomagenesis. Whereas the absence of E2F1 and E2F3 function has no impact on Myc-mediated tumor development, the absence of E2F2 substantially accelerates the time of tumor onset. Conversely, tumor development is delayed by the absence of E2F4. The enhanced early onset of tumors seen in the absence of E2F2 coincides with an expansion of immature B lineage cells that are likely to be the target for Myc oncogenesis. In contrast, the absence of E2F4 mutes the response of the lineage to Myc and there is no expansion of immature B lineage cells. We also find that distinct types of tumors emerge from the Emu-myc mice, distinguished by different patterns of gene expression, and that the relative proportions of these tumor types are affected by the absence of either E2F2 or E2F4. From these results, we conclude that there are several populations of tumors that arise from the Emu-myc model, reflecting distinct populations of cells that are susceptible to Myc-mediated oncogenesis and that the proportion of these cell populations is affected by the presence or absence of E2F activities.