Merck
  • Synergistic apoptotic effects in cancer cells by the combination of CLK and Bcl-2 family inhibitors.
Synergistic apoptotic effects in cancer cells by the combination of CLK and Bcl-2 family inhibitors.
PloS one (2020-10-17)
Aiko Murai, Shunsuke Ebara, Satoshi Sasaki, Tomohiro Ohashi, Tohru Miyazaki, Toshiyuki Nomura, Shinsuke Araki
ABSTRACT
Emerging evidence indicates that alternative splicing plays a critical role in cancer progression through abnormal expression or mutation of splicing factors. Small-molecule splicing modulators have recently attracted considerable attention as a novel class of cancer therapeutics. CDC-like kinases (CLKs) are central to exon recognition in mRNA splicing and CLK inhibitors exhibit anti-tumour activities. Most importantly, molecular mechanism-based combination strategies for cancer therapy must be considered. However, it remains unclear whether CLK inhibitors modulate expression and splicing of apoptosis-related genes, and whether CLK inhibitors enhance cytotoxicity in combination with apoptosis inducers. Here we report an appropriate mechanism-based drug combination approach. Unexpectedly, we found that the CLK inhibitor T3 rapidly induced apoptosis in A2780 cells and G2/M cell cycle arrest in HCT116 cells. Regardless of the different phenotypes of the two cancer cell types, T3 decreased the levels of anti-apoptotic proteins (cIAP1, cIAP2, XIAP, cFLIP and Mcl-1) for a short period of exposure and altered the splicing of the anti-apoptotic MCL1L and CFLAR isoform in A2780 and HCT116 cells. In contrast, other members of the Bcl-2 family (i.e., Bcl-xL and Bcl-2) were resistant to T3-induced expression and splicing modulation. T3 and a Bcl-xL/Bcl-2 inhibitor synergistically induced apoptosis. Taken together, the use of a CLK inhibitor is a novel therapeutic approach to sensitise cancer cells to Bcl-xL/Bcl-2 inhibitors.