Merck
  • Effects of Cooling Rate during Quenching and Tempering Conditions on Microstructures and Mechanical Properties of Carbon Steel Flange.
Effects of Cooling Rate during Quenching and Tempering Conditions on Microstructures and Mechanical Properties of Carbon Steel Flange.
Materials (Basel, Switzerland) (2020-09-25)
Haeju Jo, Moonseok Kang, Geon-Woo Park, Byung-Jun Kim, Chang Yong Choi, Hee Sang Park, Sunmi Shin, Wookjin Lee, Yong-Sik Ahn, Jong Bae Jeon
ABSTRACT
This study investigated the mechanical properties of steel in flanges, with the goal of obtaining high strength and high toughness. Quenching was applied alone or in combination with tempering at one of nine combinations of three temperatures TTEM and durations tTEM. Cooling rates at various flange locations during quenching were first estimated using finite element method simulation, and the three locations were selected for mechanical testing in terms of cooling rate. Microstructures of specimens were observed at each condition. Tensile test and hardness test were performed at room temperature, and a Charpy impact test was performed at -46 °C. All specimens had a multiphase microstructure composed of matrix and secondary phases, which decomposed under the various tempering conditions. Decrease in cooling rate (CR) during quenching caused reduction in hardness and strength but did not affect low-temperature toughness significantly. After tempering, hardness and strength were reduced and low-temperature toughness was increased. Microstructures and mechanical properties under the various tempering conditions and CRs during quenching were discussed. This work was based on the properties directly obtained from flanges under industrial processes and is thus expected to be useful for practical applications.
Sigma-Aldrich
(R)-Pantetheine, ≥95.0% (HPLC)