Merck
  • Autophagy under glucose starvation enhances protein translation initiation in response to re-addition of glucose in C2C12 myotubes.
Autophagy under glucose starvation enhances protein translation initiation in response to re-addition of glucose in C2C12 myotubes.
FEBS open bio (2020-09-04)
Naoya Nakai, Saki Kitai, Noriko Iida, Sachika Inoue, Kazuhiko Higashida
ABSTRACT
Proteolysis is known to play a crucial role in maintaining skeletal muscle mass and function. Autophagy is a conserved intracellular process for the bulk degradation of proteins in lysosomes. Although nutrient starvation is known to induce autophagy, the effect of nutrient repletion following starvation on the mTOR pathway-mediated protein translation remains unclear. In the present study, we examined the effect of glucose starvation on the initiation of protein translation in response to glucose re-addition in C2C12 myotubes. Glucose starvation decreased the phosphorylation of p70 S6 kinase (p70S6K), a bonafide marker for protein translation initiation. Following re-addition of glucose, phosphorylation of p70S6K markedly increased only in glucose-starved cells. Inhibiting autophagy using pharmacological inhibitors diminished the effect of glucose re-addition on the phosphorylation of p70S6K, whereas inhibition of the ubiquitin-proteasome system did not exert any effect. In conclusion, autophagy under glucose starvation partially accounts for the activation of translation initiation by re-addition of glucose.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Penicillin–Streptomycin Solution Hybri-Max, with 10,000 units penicillin and 10mg/ml streptomycin in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for hybridoma
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma