Merck
  • Efficacy of a Semi Automated Commercial Closed System for Autologous Leukocyte- and Platelet-Rich Plasma (l-prp) Production in Dogs: A Preliminary Study.
Efficacy of a Semi Automated Commercial Closed System for Autologous Leukocyte- and Platelet-Rich Plasma (l-prp) Production in Dogs: A Preliminary Study.
Animals : an open access journal from MDPI (2020-08-08)
Roberta Perego, Eva Spada, Luciana Baggiani, Piera Anna Martino, Daniela Proverbio
ABSTRACT
To characterize the cellular composition (platelets, erythrocytes, and leukocytes) and determine platelet-derived growth factor isoform BB (PDGF-BB) concentration in canine leukocyte- and platelet rich plasma (L-PRP) produced using a commercial semi-automated closed system. Twenty milliliters of citrated whole blood were obtained from 30 healthy un-sedated canine blood donors and processed using a semi-automated completely closed commercial system (CPUNT 20, Eltek group, Casale Monferrato, Alessandria, Italy) according to the manufacturer's instructions. Erythrocyte, leukocyte, and platelet counts were determined in both whole blood (WB) and resultant L-PRP. The PDGF-BB concentration was evaluated after bovine thrombin activation of 10 L-PRP samples. This commercial system produced on average 2.3 ± 0.7 mL of L-PRP containing a high concentration of platelets (767,633 ± 291,001 μL, p < 0.001), with a 4.4 fold increase in platelet count, lower concentration of erythrocytes (528,600 ± 222,773 μL, p < 0.001) and similar concentration of leukocytes (8422 ± 6346 μL, p = 0.9918) compared with WB. L-PRP had an average of 3442 ± 2061 pg/mL of PDGF-BB after thrombin activation. Neutrophils, lymphocytes and monocytes average percent content in L-PRP was 14.8 ± 13.2, 71.7 ± 18.5 and 10.7 ± 6.4, respectively. Sterile canine L-PRP prepared using this semi-automated closed system is easy to obtain, produces a significant increase in platelet count compared to WB and contains a detectable concentration of PDGF-BB after activation. Additional in vitro and in vivo studies are needed to assess inflammatory markers concentration and the therapeutic efficacy of this L-PRP in dogs.
Sigma-Aldrich
Human PLG (Plasminogen) ELISA Kit, for cell culture supernatants, plasma, and serum samples