Merck
  • Potentiation of catechin gallate-mediated sensitization of Staphylococcus aureus to oxacillin by nongalloylated catechins.
Potentiation of catechin gallate-mediated sensitization of Staphylococcus aureus to oxacillin by nongalloylated catechins.
Antimicrobial agents and chemotherapy (2006-01-27)
Paul D Stapleton, Saroj Shah, Yukihiko Hara, Peter W Taylor
ABSTRACT
(-)-Epicatechin gallate (ECg) and (-)-epigallocatechin gallate (EGCg) reduce oxacillin resistance in mecA-containing strains of Staphylococcus aureus. Their binding to staphylococcal cells is enhanced by the nongalloyl analogues (-)-epicatechin (EC) and (-)-epigallocatechin (EGC). EC and EGC significantly increased the capacity of ECg and EGCg to reduce levels of staphylococcal oxacillin resistance.
Sigma-Aldrich
(−)-Catechin gallate, ≥98% (HPLC), from green tea