- Complete Genome Sequence of Staphylococcus aureus Strain Wood 46.
Complete Genome Sequence of Staphylococcus aureus Strain Wood 46.
Genome announcements (2017-04-01)
Manasi Balachandran, Matthew C Riley, David A Bemis, Stephen A Kania
PMID28360163
ABSTRACT
Here, we report the first complete genome sequence of the Staphylococcus aureus strain Wood 46. Wood 46 has played an important role in understanding the virulence and pathogenesis of S. aureus infections. This report will assist efforts in vaccine development against methicillin-resistant S. aureus (MRSA) infections.