Merck
Effects of UV Stabilizers on Polypropylene Outdoors.
Materials (Basel, Switzerland) (2020-04-05)
Witold Brostow, Xinyao Lu, Osman Gencel, Allison T Osmanson
ABSTRACT
Hindered amine light stabilizers (HALSs) and nano ZnO were used to stabilize polypropylene (PP) film-based formulations that were exposed to ultraviolet (UV) light for different lengths of time, simulating the harsh outdoor weather of Dallas, Texas, USA. UV doses applied in our laboratory are 121 times larger than the UV dose provided by the sunlight in Texas. 15 different compositions were studied. Tensile behavior, UV transmittance, thermal stability (by thermogravimetric analysis) and dynamic friction of the so exposed PP-based films were determined. Scanning electron micrographs of fracture surfaces were obtained. Nano-ZnO-containing stabilizers impart strong UV resistance to our films. The combination of HALSs and nano-ZnO stabilizers makes the PP films harder-which is important for some PP applications, such as toy manufacturing.
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)