Merck
  • Racemic total synthesis and evaluation of the biological activities of the isoquinoline-benzylisoquinoline alkaloid muraricine.
Racemic total synthesis and evaluation of the biological activities of the isoquinoline-benzylisoquinoline alkaloid muraricine.
Archiv der Pharmazie (2020-05-26)
Ramona Schütz, Martin Müller, Susanne Gerndt, Karin Bartel, Franz Bracher
ABSTRACT
The first racemic total synthesis of the isoquinoline-benzylisoquinoline alkaloid muraricine is reported herein. Pharmacological characterization identified muraricine as a moderate inhibitor of P-glycoprotein, a crucial factor of multidrug resistance in cancer. When combined with vincristine, muraricine partly reversed the chemoresistance of vincristine-resistant leukemia cells at a nontoxic concentration. Furthermore, no cytotoxic effects on noncancerous human cells in therapeutically relevant concentrations were observed.
Sigma-Aldrich
TPC2-A1-N, ≥98% (HPLC)