Merck
  • CTP synthase polymerization in germline cells of the developing Drosophila egg supports egg production.
CTP synthase polymerization in germline cells of the developing Drosophila egg supports egg production.
Biology open (2020-06-26)
Jacqueline C Simonet, Maya J Foster, Eric M Lynch, Justin M Kollman, Emmanuelle Nicholas, Alana M O'Reilly, Jeffrey R Peterson
ABSTRACT
Polymerization of metabolic enzymes into micron-scale assemblies is an emerging mechanism for regulating their activity. CTP synthase (CTPS) is an essential enzyme in the biosynthesis of the nucleotide CTP and undergoes regulated and reversible assembly into large filamentous structures in organisms from bacteria to humans. The purpose of these assemblies is unclear. A major challenge to addressing this question has been the inability to abolish assembly without eliminating CTPS protein. Here we demonstrate that a recently reported point mutant in CTPS, Histidine 355A (H355A), prevents CTPS filament assembly in vivo and dominantly inhibits the assembly of endogenous wild-type CTPS in the Drosophila ovary. Expressing this mutant in ovarian germline cells, we show that disruption of CTPS assembly in early stage egg chambers reduces egg production. This effect is exacerbated in flies fed the glutamine antagonist 6-diazo-5-oxo-L-norleucine, which inhibits de novo CTP synthesis. These findings introduce a general approach to blocking the assembly of polymerizing enzymes without eliminating their catalytic activity and demonstrate a role for CTPS assembly in supporting egg production, particularly under conditions of limited glutamine metabolism.This article has an associated First Person interview with the first author of the paper.
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%