Merck
  • Eriodictyol-7-O-glucoside, a novel Nrf2 activator, confers protection against cisplatin-induced toxicity.
Eriodictyol-7-O-glucoside, a novel Nrf2 activator, confers protection against cisplatin-induced toxicity.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-04-03)
Qingwen Hu, Donna D Zhang, Limei Wang, Hongxiang Lou, Dongmei Ren
ABSTRACT
Eriodictyol-7-O-glucoside, a flavonoid isolated from Dracocephalum rupestre, is among the most potent free radical scavenger. In the present study, we identified eriodictyol-7-O-glucoside as a novel nuclear factor E2-related factor 2 (Nrf2) activator using a high-throughput cellular screening method. This compound activated Nrf2 signaling pathway and was able to stabilize Nrf2 by delaying Nrf2 degradation, resulting in accumulation of Nrf2 protein and activation of the Nrf2-dependent protective response. Recent studies have suggested that activation of Nrf2 pathway would confer protection against cisplatin-induced toxicity. The protective role of eriodictyol-7-O-glucoside in cisplatin-induced toxicity was investigated in a human renal mesangial cell line, HRMC. Cotreatment of HRMC cells with eriodictyol-7-O-glucoside significantly improved cell survival under cisplatin exposure. These findings demonstrated the feasibility of using natural compounds targeting Nrf2 as a therapeutic approach to subvert the side effects of cisplatin in normal cells.