Merck
  • Discovery of potent and selective inhibitors of human platelet-type 12- lipoxygenase.
Discovery of potent and selective inhibitors of human platelet-type 12- lipoxygenase.
Journal of medicinal chemistry (2011-07-12)
Victor Kenyon, Ganesha Rai, Ajit Jadhav, Lena Schultz, Michelle Armstrong, J Brian Jameson, Steven Perry, Netra Joshi, James M Bougie, William Leister, David A Taylor-Fishwick, Jerry L Nadler, Michael Holinstat, Anton Simeonov, David J Maloney, Theodore R Holman
ABSTRACT
We report the discovery of novel small molecule inhibitors of platelet-type 12-human lipoxygenase, which display nanomolar activity against the purified enzyme, using a quantitative high-throughput screen (qHTS) on a library of 153607 compounds. These compounds also exhibit excellent specificity, >50-fold selectivity vs the paralogues, 5-human lipoxygenase, reticulocyte 15-human lipoxygenase type-1, and epithelial 15-human lipoxygenase type-2, and >100-fold selectivity vs ovine cyclooxygenase-1 and human cyclooxygenase-2. Kinetic experiments indicate this chemotype is a noncompetitive inhibitor that does not reduce the active site iron. Moreover, chiral HPLC separation of two of the racemic lead molecules revealed a strong preference for the (-)-enantiomers (IC(50) of 0.43 ± 0.04 and 0.38 ± 0.05 μM) compared to the (+)-enantiomers (IC(50) of >25 μM for both), indicating a fine degree of selectivity in the active site due to chiral geometry. In addition, these compounds demonstrate efficacy in cellular models, which underscores their relevance to disease modification.
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Sigma-Aldrich
Baicalein, 98%