Merck
  • LncRNA Gm14205 induces astrocytic NLRP3 inflammasome activation via inhibiting oxytocin receptor in postpartum depression.
LncRNA Gm14205 induces astrocytic NLRP3 inflammasome activation via inhibiting oxytocin receptor in postpartum depression.
Bioscience reports (2020-07-25)
Jialei Zhu, Jing Tang
ABSTRACT
Postpartum depression (PPD) is a kind of mental disorder characterized by persistent low emotions in puerperium. The most significant physiological change in postpartum is lactation which is regulated by oxytocin receptor (OXTR). However, whether OXTR is related to pathological process of PPD and the potential mechanism still remain unclear. In the present study, we prepared hormone-simulated pregnancy (HSP)-induced PPD mouse model and found that the protein level of OXTR in hippocampus of PPD model mice was down-regulated and Nod-like receptor protein 3 (NLRP3) inflammasome was activated. We identified five long non-coding RNAs (lncRNAs) related to PPD by transcriptome sequencing, including three up-regulated and two down-regulated. The five lncRNAs were associated with the signaling pathway of OXTR according to the bioinformatics analysis. Furthermore, we focused on one of the five lncRNAs, Gm14205, and found that it targeted OXTR which inhibited astrocytic NLRP3 inflammasome activation in hippocampal primary astrocytes. These findings illustrate that OXTR has protective effects in PPD by inhibiting NLRP3 inflammasome activation and provides a new strategy for targeting lncRNA Gm14205 in the pathogenesis of PPD.