Merck
  • Fabrication of Perfusable Vascular Channels and Capillaries in 3D Liver-like Tissue.
Fabrication of Perfusable Vascular Channels and Capillaries in 3D Liver-like Tissue.
Scientific reports (2020-04-15)
Nobuhito Mori, Yuka Akagi, Yukiko Imai, Yuzo Takayama, Yasuyuki S Kida
ABSTRACT
Although various production methods for 3D vascularised tissues have been developed, constructing capillary-like structures branching from perfusable large channels remains difficult. This study describes a method to fabricate tube-shaped 3D liver-like tissue (tubular liver tissue) with large channels and capillary-like structures using a perfusion device. The perfusion device functions as an interface between the tissue and an external pump, as it has connectors equipped with anchors that hold the tissue in response to its shrinkage, which is accompanied by the self-organisation of capillary-like structures. Histological analysis revealed that perfusion via the large channel induced capillary formation around the channel and maintained proper tissue functions. Accompanied by structural examinations, global gene expression analysis supported this finding; specifically, genes involved in angiogenesis were enriched in the perfused condition. Furthermore, we confirmed the penetrability of the capillary-like structures by infusing India ink, as well as substance exchange by measuring the amounts of secreted albumin. These lines of evidence indicate that our method can be used to construct 3D tissues, which is useful for fields of in vitro tissue regeneration for drug development and regenerative medicine.