Merck
  • Novel polymer-supported 2-(diphenylmethylsilyl)ethoxymethyl chloride (DSEM-Cl) linker.
Novel polymer-supported 2-(diphenylmethylsilyl)ethoxymethyl chloride (DSEM-Cl) linker.
Chemical communications (Cambridge, England) (2002-09-21)
K Kim, B Wang
ABSTRACT
A synthetic method to prepare a novel polymer-supported 2-(diphenylmethylsilyl)ethoxymethyl chloride (DSEM-Cl) linker and its applications are described.
Sigma-Aldrich
2-(Trimethylsilyl)ethoxymethyl chloride, ≥95.0% (GC)