Merck
PLGA-based nanoparticles: an overview of biomedical applications.
Journal of controlled release : official journal of the Controlled Release Society (2012-02-23)
Fabienne Danhier, Eduardo Ansorena, Joana M Silva, Régis Coco, Aude Le Breton, Véronique Préat
ABSTRACT
Poly(lactic-co-glycolic acid) (PLGA) is one of the most successfully developed biodegradable polymers. Among the different polymers developed to formulate polymeric nanoparticles, PLGA has attracted considerable attention due to its attractive properties: (i) biodegradability and biocompatibility, (ii) FDA and European Medicine Agency approval in drug delivery systems for parenteral administration, (iii) well described formulations and methods of production adapted to various types of drugs e.g. hydrophilic or hydrophobic small molecules or macromolecules, (iv) protection of drug from degradation, (v) possibility of sustained release, (vi) possibility to modify surface properties to provide stealthness and/or better interaction with biological materials and (vii) possibility to target nanoparticles to specific organs or cells. This review presents why PLGA has been chosen to design nanoparticles as drug delivery systems in various biomedical applications such as vaccination, cancer, inflammation and other diseases. This review focuses on the understanding of specific characteristics exploited by PLGA-based nanoparticles to target a specific organ or tissue or specific cells.
Sigma-Aldrich
NanoFabTx PEG-PLGA drug formulation screening kit, for synthesis of PEGylated nanoparticles
Sigma-Aldrich
NanoFabTx PLGA-nano, for synthesis of 100 and 200 nm particles