Merck
  • Novel phosphorus-containing 17beta-side chain mifepristone analogues as progesterone receptor antagonists.
Novel phosphorus-containing 17beta-side chain mifepristone analogues as progesterone receptor antagonists.
Steroids (2006-08-29)
Weiqin Jiang, George Allan, Xin Chen, James J Fiordeliso, Olivia Linton, Pamela Tannenbaum, Jun Xu, Peifang Zhu, Joseph Gunnet, Keith Demarest, Scott Lundeen, Zhihua Sui
ABSTRACT
A novel series of steroidal compounds were designed and synthesized with various phosphorus-containing groups on the 17beta-side chain as progesterone receptor antagonists. The structure-activity relationships of these compounds are discussed. Selected compounds were tested in an rat progesterone-sensitive assay. Some of these compounds are more potent than mifepristone, with a better selectivity profile in differentiating progesterone receptor from glucocorticoid receptor.