Merck
  • Cis- and trans-regulations of pre-mRNA splicing by RNA editing enzymes influence cancer development.
Cis- and trans-regulations of pre-mRNA splicing by RNA editing enzymes influence cancer development.
Nature communications (2020-02-09)
Sze Jing Tang, Haoqing Shen, Omer An, HuiQi Hong, Jia Li, Yangyang Song, Jian Han, Daryl Jin Tai Tay, Vanessa Hui En Ng, Fernando Bellido Molias, Ka Wai Leong, Priyankaa Pitcheshwar, Henry Yang, Leilei Chen
ABSTRACT
RNA editing and splicing are the two major processes that dynamically regulate human transcriptome diversity. Despite growing evidence of crosstalk between RNA editing enzymes (mainly ADAR1) and splicing machineries, detailed mechanistic explanations and their biological importance in diseases, such as cancer are still lacking. Herein, we identify approximately a hundred high-confidence splicing events altered by ADAR1 and/or ADAR2, and ADAR1 or ADAR2 protein can regulate cassette exons in both directions. We unravel a binding tendency of ADARs to dsRNAs that involves GA-rich sequences for editing and splicing regulation. ADAR1 edits an intronic splicing silencer, leading to recruitment of SRSF7 and repression of exon inclusion. We also present a mechanism through which ADAR2 binds to dsRNA formed between GA-rich sequences and polypyrimidine (Py)-tract and precludes access of U2AF65 to 3' splice site. Furthermore, we find these ADARs-regulated splicing changes per se influence tumorigenesis, not merely byproducts of ADARs editing and binding.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution