Merck
  • Size-Dependent Thermo- and Photoresponsive Plasmonic Properties of Liquid Crystalline Gold Nanoparticles.
Size-Dependent Thermo- and Photoresponsive Plasmonic Properties of Liquid Crystalline Gold Nanoparticles.
Materials (Basel, Switzerland) (2020-02-23)
Aleksander Promiński, Ewelina Tomczyk, Mateusz Pawlak, Agnieszka Jędrych, Józef Mieczkowski, Wiktor Lewandowski, Michał Wójcik
ABSTRACT
Achieving remotely controlled, reversibly reconfigurable assemblies of plasmonic nanoparticles is a prerequisite for the development of future photonic technologies. Here, we obtained a series of gold-nanoparticle-based materials which exhibit long-range order, and which are controlled with light or thermal stimuli. The influence of the metallic core size and organic shell composition on the switchability is considered, with emphasis on achieving light-responsive behavior at room temperature and high yield production of nanoparticles. The latter translates to a wide size distribution of metallic cores but does not prevent their assembly into various, switchable 3D and 2D long-range ordered structures. These results provide clear guidelines as to the impact of size, size distribution, and organic shell composition on self-assembly, thus enhancing the smart design process of multi-responsive nanomaterials in a condensed state, hardly attainable by other self-assembly methods which usually require solvents.
Sigma-Aldrich
Diisopropyl azodicarboxylate, 98%
Sigma-Aldrich
4-(Chloromethyl)benzoyl chloride, 97%