Merck
  • Wilms' tumour 1 (WT1) negatively regulates the expression of connexin 43 via a non-canonical Wnt signalling pathway in cultured bovine Sertoli cells.
Wilms' tumour 1 (WT1) negatively regulates the expression of connexin 43 via a non-canonical Wnt signalling pathway in cultured bovine Sertoli cells.
Reproduction, fertility, and development (2020-02-06)
Xue Wang, Ziming Wang, S O Adeniran, Fushuo Huang, Mingjun Ma, Han Zhang, Xiaoyu Li, Peng Zheng, Guixue Zhang
ABSTRACT
The gap junction protein connexin (Cx) 43 between adjacent Sertoli cells (SCs) is the main testicular factor regulating the growth and development of SCs, and plays a vital role in controlling cell differentiation and maturation. However, the endogenous testicular factors that regulate Cx43 and the downstream signalling pathways that mediate Cx43-dependent SC differentiation are unclear. In this study, high-purity SCs were isolated from newborn calves' testes by differential adherence. The SCs were then cultured invitro and treated with short interference RNA to knockdown endogenous Wilms' tumour 1 (WT1). In WT1-knockdown SCs, Cx43 expression was upregulated. To elucidate the intracellular signalling mechanism of Cx43 in the differentiation and maturation of immature SCs, SCs were treated simultaneously with non-canonical Wnt signalling inhibitors CCG-1423 and GO-6983; in these SCs, Cx43 expression was upregulated. Together, these data indicate that WT1 negatively regulates the expression of Cx43 produced from SCs via a non-canonical Wnt signalling pathway in cultured bovine SCs.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
MISSION® esiRNA, targeting human WT1
Sigma-Aldrich
Triton X-100, laboratory grade