Merck
  • Carbohydrate carbonyl mobility--the key process in the formation of alpha-dicarbonyl intermediates.
Carbohydrate carbonyl mobility--the key process in the formation of alpha-dicarbonyl intermediates.
Carbohydrate research (2004-06-09)
Oliver Reihl, Thorsten M Rothenbacher, Markus O Lederer, Wolfgang Schwack
ABSTRACT
Covalently cross-linked proteins are among the major modifications caused by the advanced Maillard reaction. In the present study, the formation pathway of the dideoxyosone N6-(2,3-dihydroxy-5,6-dioxohexyl)-L-lysine is shown. To elucidate the formation of this glucose-derived dideoxyosone D-lactose (O-beta-D-galp-(1-->4)-D-glcp) and D-glucose-6-phosphate were incubated with lysine in the presence of the trapping reagent o-phenylenediamine (OPD). Synthesis and unequivocal structural characterization were reported for the quinoxalines of the dideoxyosones N6-(5,6-dihydroxy-2,3-dioxohexyl)-L-lysine and N6-(2,3-dihydroxy-4,5-dioxohexyl)-L-lysine, respectively. Additionally, dicarbonyl compounds derived from D-erythrose, D-glycero-D-mannoheptose, and D-gluco-L-talooctose were synthesized and structurally characterized.
Sigma-Aldrich
D-(−)-Erythrose, ≥75% (TLC), syrup