Merck
  • A serine/threonine-specific protein kinase of Haemonchus contortus with a role in the development.
A serine/threonine-specific protein kinase of Haemonchus contortus with a role in the development.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-01-08)
Wenda Di, Robin B Gasser, Li He, Fangfang Li, Xiaofang Liu, Caixian Zhou, Yanqin Zhou, Rui Fang, Junlong Zhao, Min Hu
ABSTRACT
In the free-living nematode Caenorhabditis elegans, the serine/threonine-specific protein kinase, AKT, is known to play a key role in dauer formation, life-span, and stress-resistance through the insulin-like signaling pathway. Although the structure and function of AKT-coding genes of C. elegans are understood, this is not the case for homologous genes in parasitic nematodes. In the present study, we explored a C. elegans akt-1 gene homolog in the parasitic nematode Haemonchus contortus, investigated its transcript isoforms (Hc-akt-1a and Hc-akt-1b), and studied expression and function using both homologous and heterologous functional genomic tools. In C. elegans, we showed that the predicted promoter of Hc-akt-1 drives substantial expression in ASJ neurons of the N2 (wild-type) strain. In H. contortus (Haecon-5 stain), RNAi (soaking) led to a significantly decreased transcript abundance for both Hc-akt-1a and Hc-akt-1b, and reduced larval development in larval stages in vitro. Chemical inhibition was also shown to block larval development. Taken together, the evidence from this study points to a key functional role for Hc-akt-1 in H. contortus.
Sigma-Aldrich
5-(2-Benzothiazolyl)-3-ethyl-2-[2-(methylphenylamino)ethenyl]-1-phenyl-1H-benzimidazolium iodide, ≥98% (HPLC)