- [Synthesis of oligosaccharide determinants with amide spacers of T-type antigens].
[Synthesis of oligosaccharide determinants with amide spacers of T-type antigens].
Carbohydrate research (1982-06-16)
H Paulsen, J C Jacquinet, W Rust
PMID6179620
ABSTRACT
The hapten of the T-antigen was synthesized with a peptide-like amide-spacer as 2-(4-methoxycarbonylbutanecarboxamido)ethyl 2-acetamido-2-deoxy-3-O-beta-D-galactopyranosyl-alpha-D-galactopyranoside and coupled with serum albumin to give a synthetic antigen. Other O-beta-D-galactopyranosyl haptens, 2-(4-methoxycarbonylbutanecarboxamido)ethyl 2-acetamido-2-deoxy-4-O-beta-D-galactopyranosyl-alpha-D-galactopyranoside, O-beta-D-galactopyranosyl-(1 leads to 3)-O-[beta-D-galactopyranosyl-(1 leads to 4)]-2-acetamido-2-deoxy-alpha-D-galactopyranoside, and 2-acetamido-2-deoxy-3-O-beta-D-galactopyranosyl-alpha-D-glucopyranoside, the last compound being the determinant of the Lewis Lec antigen, were also synthesized.