Merck
  • Effects of a cosmetic 'anti-ageing' product improves photoaged skin [corrected].
Effects of a cosmetic 'anti-ageing' product improves photoaged skin [corrected].
The British journal of dermatology (2009-05-15)
R E B Watson, S Ogden, L F Cotterell, J J Bowden, J Y Bastrilles, S P Long, C E M Griffiths
ABSTRACT
Very few over-the-counter cosmetic 'anti-ageing' products have been subjected to a rigorous double-blind, vehicle-controlled trial of efficacy. Previously we have shown that application of a cosmetic 'anti-ageing' product to photoaged skin under occlusion for 12 days can stimulate the deposition of fibrillin-1. This observation infers potential to repair and perhaps clinically improve photoaged skin. We examined another similar over-the-counter cosmetic 'anti-ageing' product using both the patch test assay and a 6-month double-blind, randomized controlled trial (RCT), with a further 6-month open phase to assess clinical efficacy in photoaged skin. For the patch test, commercially [corrected] available test product and its vehicle were applied occluded for 12-days to photoaged forearm skin (n = 10) prior to biopsy and immunohistochemical assessment of fibrillin-1; all-transretinoic acid (RA) [corrected] was used as a positive control. Sixty photoaged subjects were recruited to the RCT (test product, n = 30 vs. vehicle, n = 30; once daily for 6-months; face & hands) [corrected] with clinical assessments performed at recruitment and following 1-, 3- & 6-months of use [corrected]. Twenty-eight subjects had skin biopsies (dorsal wrist) at baseline and at 6 months of treatment for immunohistochemical assessment of fibrillin-1 (test product, n = 15; vehicle, n = 13). All subjects [corrected] received test product for a further 6-months. Final clinical assessments were performed at the end of this open period; 27 subjects received test product for 12-months [corrected]. In the 12-day patch test assay, we observed significant immunohistological deposition of fibrillin-1 in skin treated by test product and RA as compared to untreated baseline (P = 0.005 and 0.015 respectively). In the clinical RCT, at 6 months, compared to baseline assessment, 43% of subjects on test product had an improvement in facial wrinkles (P = 0.013), whereas only 22% of subjects using vehicle had clinical improvement (P = ns). Between group comparison of test product and vehicle was non-significant (P = 0.10). After 12 months, there was a significant benefit of test product over that projected for vehicle (70% vs. 33% of subjects improving; combined Wilcoxon rank tests, P = 0.026). There was significant deposition of fibrillin-1 in skin treated for 6 months with test product (mean +/- SE; vehicle, 1.84 +/- 0.23; test product, 2.57 +/- 0.19; P = 0.019). An over-the-counter cosmetic 'anti-ageing' product demonstrated clear benefit over vehicle in fibrillin-1 deposition over a 6-month trial period. There was a corresponding but non-significant trend towards clinical improvement in facial wrinkles. Clinical improvements in the treated group were increased after a further 6-months of use. This study demonstrates that a cosmetic may improve the appearance of wrinkles and further supports the use of fibrillin-1 as a robust biomarker for repair of photoaged dermis.