Merck
  • Cyclin B1/Cdk1 binds and phosphorylates Filamin A and regulates its ability to cross-link actin.
Cyclin B1/Cdk1 binds and phosphorylates Filamin A and regulates its ability to cross-link actin.
FEBS letters (2007-04-06)
I Howard Cukier, Yun Li, Jonathan M Lee
ABSTRACT
Substantial actin remodelling occurs prior to mitosis as cells alter their shape in preparation for cytokinesis. In mammalian cells, mitosis is initiated by a heterodimer of cyclin B1 and the cyclin dependent kinase 1 (Cdk1) serine/threonine kinase. In this report. we show that human cyclin B1 binds the actin cross-linking protein Filamin-A (FLNa). The proteins co-immunoprecipitate and co-localize in mitotic human cells. We find that cyclin B1/Cdk1 can phosphorylate FLNa in vitro and reduce its ability to gelate actin. We have also identified serine 1436 as one FLNa residue phosphorylated by cyclin B1/Cdk1 in vitro. Our results suggest a role for cyclin B1/Cdk1 in FLNa-dependent actin remodelling.