Merck
  • The noncoding RNAs SNORD50A and SNORD50B bind K-Ras and are recurrently deleted in human cancer.
The noncoding RNAs SNORD50A and SNORD50B bind K-Ras and are recurrently deleted in human cancer.
Nature genetics (2015-11-26)
Zurab Siprashvili, Dan E Webster, Danielle Johnston, Rajani M Shenoy, Alexander J Ungewickell, Aparna Bhaduri, Ross Flockhart, Brian J Zarnegar, Yonglu Che, Francesca Meschi, Joseph D Puglisi, Paul A Khavari
ABSTRACT
Small nucleolar RNAs (snoRNAs) are conserved noncoding RNAs best studied as ribonucleoprotein (RNP) guides in RNA modification. To explore their role in cancer, we compared 5,473 tumor-normal genome pairs to identify snoRNAs with frequent copy number loss. The SNORD50A-SNORD50B snoRNA locus was deleted in 10-40% of 12 common cancers, where its loss was associated with reduced survival. A human protein microarray screen identified direct SNORD50A and SNORD50B RNA binding to K-Ras. Loss of SNORD50A and SNORD50B increased the amount of GTP-bound, active K-Ras and hyperactivated Ras-ERK1/ERK2 signaling. Loss of these snoRNAs also increased binding by farnesyltransferase to K-Ras and increased K-Ras prenylation, suggesting that KRAS mutation might synergize with SNORD50A and SNORD50B loss in cancer. In agreement with this hypothesis, CRISPR-mediated deletion of SNORD50A and SNORD50B in KRAS-mutant tumor cells enhanced tumorigenesis, and SNORD50A and SNORD50B deletion and oncogenic KRAS mutation co-occurred significantly in multiple human tumor types. SNORD50A and SNORD50B snoRNAs thus directly bind and inhibit K-Ras and are recurrently deleted in human cancer.