Merck
  • Systemic HIV-1 infection produces a unique glial footprint in humanized mouse brains.
Systemic HIV-1 infection produces a unique glial footprint in humanized mouse brains.
Disease models & mechanisms (2017-11-01)
Weizhe Li, Santhi Gorantla, Howard E Gendelman, Larisa Y Poluektova
ABSTRACT
Studies of innate glial cell responses for progressive human immunodeficiency virus type one (HIV-1) infection are limited by a dearth of human disease-relevant small-animal models. To overcome this obstacle, newborn NOD/SCID/IL2Rγc-/- (NSG) mice were reconstituted with a humanized brain and immune system. NSG animals of both sexes were transplanted with human neuroglial progenitor cells (NPCs) and hematopoietic stem cells. Intraventricular injection of NPCs symmetrically repopulated the mouse brain parenchyma with human astrocytes and oligodendrocytes. Human glia were in periventricular areas, white matter tracts, the olfactory bulb and the brain stem. HIV-1 infection led to meningeal and perivascular human leukocyte infiltration into the brain. Species-specific viral-neuroimmune interactions were identified by deep RNA sequencing. In the corpus callosum and hippocampus of infected animals, overlapping human-specific transcriptional alterations for interferon type 1 and 2 signaling pathways (STAT1, STAT2, IRF9, ISG15, IFI6) and a range of host antiviral responses (MX1, OAS1, RSAD2, BST2, SAMHD1) were observed. Glial cytoskeleton reorganization, oligodendrocyte differentiation and myelin ensheathment (MBP, MOBP, PLP1, MAG, ZNF488) were downregulated. The data sets were confirmed by real-time PCR. These viral defense-signaling patterns paralleled neuroimmune communication networks seen in HIV-1-infected human brains. In this manner, this new mouse model of neuroAIDS can facilitate diagnostic, therapeutic and viral eradication strategies for an infected nervous system.