Merck
  • Dynamics of γ-tubulin cytoskeleton in HL-60 leukemia cells undergoing differentiation and apoptosis by all-trans retinoic acid.
Dynamics of γ-tubulin cytoskeleton in HL-60 leukemia cells undergoing differentiation and apoptosis by all-trans retinoic acid.
Molecular medicine reports (2011-10-21)
Ahmad Shariftabrizi, Shahin Ahmadian, Yaghub Pazhang
ABSTRACT
Microtubules are important components of the cell cytoskeleton, participating in protein localization and cell signaling. The capacity of leukemia cells to re-organize their microtubules is considered an integral part of differentiation in these cells in order to become mature granulocytes through treatment with all-trans retinoic acid (ATRA), an established drug for treating acute promyelocytic leukemia. In this study we examined γ-, α- and acetylated-α-tubulin content, their patterns of distribution in the cytoplasm, and the potency of centrosomes in re-organizing microtubules in different stages of ATRA-induced differentiation and apoptosis of the HL-60 cell line. The γ-tubulin content was dramatically increased following differentiation of HL-60 cells, and was then decreased after apoptosis. We also found that γ-tubulin had a diffuse, cytoplasmic pattern following apoptosis compared to the focal, centrosomal accumulation of γ-tubulin in differentiated cells. Differentiated cells had the ability to re-organize their microtubule network following nocodazole challenge testing, whereas undifferentiated cells did not show a similar ability. α-tubulin was more regularly organized in differentiated cells, and did not reveal any specific pattern of polymerization in apoptotic cells. Acetylated-α-tubulin generally followed the same organization patterns after differentiation, as that which occurred for α-tubulin. Our data is suggestive of a centrosomal and organized nucleation pattern of microtubules in HL-60 cells following differentiation, possibly mediated through up-regulation of γ-tubulin.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid