Merck
  • Crosslinking activity of non-muscle myosin II is not sufficient for embryonic cytokinesis in C. elegans.
Crosslinking activity of non-muscle myosin II is not sufficient for embryonic cytokinesis in C. elegans.
Development (Cambridge, England) (2019-10-05)
Daniel S Osório, Fung-Yi Chan, Joana Saramago, Joana Leite, Ana M Silva, Ana F Sobral, Reto Gassmann, Ana Xavier Carvalho
ABSTRACT
Cytokinesis in animal cells requires the assembly and constriction of a contractile actomyosin ring. Non-muscle myosin II is essential for cytokinesis, but the role of its motor activity remains unclear. Here, we examine cytokinesis in C. elegans embryos expressing non-muscle myosin motor mutants generated by genome editing. Two non-muscle motor-dead myosins capable of binding F-actin do not support cytokinesis in the one-cell embryo, and two partially motor-impaired myosins delay cytokinesis and render rings more sensitive to reduced myosin levels. Further analysis of myosin mutants suggests that it is myosin motor activity, and not the ability of myosin to crosslink F-actin, that drives the alignment and compaction of F-actin bundles during contractile ring assembly, and that myosin motor activity sets the pace of contractile ring constriction. We conclude that myosin motor activity is required at all stages of cytokinesis. Finally, characterization of the corresponding motor mutations in C. elegans major muscle myosin shows that motor activity is required for muscle contraction but is dispensable for F-actin organization in adult muscles.This article has an associated 'The people behind the papers' interview.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Muscle acetone powder from rabbit