Merck
The tumor suppressor miR-138-5p targets PD-L1 in colorectal cancer.
Oncotarget (2016-06-02)
Lian Zhao, Haibo Yu, Shuijing Yi, Xiaowei Peng, Peng Su, Zhiming Xiao, Rui Liu, Anliu Tang, Xiayu Li, Fen Liu, Shourong Shen
ABSTRACT
microRNAs (miRNAs) play critical roles in cancer development and progression. This study investigated the effects of miR-138-5p in human colorectal cancer (CRC) development. miR-138-5p was frequently downregulated in CRC tissues and was associated with advanced clinical stage, lymph node metastasis and poor overall survival. We found that miR-138-5p decreased expression of programmed cell death ligand 1 (PD-L1) through interaction with its PD-L1 3' untranslated region. miR-138-5p also dramatically suppressed CRC cell growth in vitro and inhibited tumorigenesis in vivo. PD-L1 and miR-138-5p levels were inversely correlated in human CRC tumors, and miR-138-5p inhibited PD-L1 expression in tumor models. These results suggest that miR-138-5p is a tumor suppressor in CRC, and its effects are exerted at least partially through PD-L1 downregulation. Low miR-138-5p and high PD-L1 levels correlated with shorter overall CRC patient survival, indicating that miR-138-5p and PD-L1 may serve as CRC biomarkers for risk group assignment, optimal therapy selection and clinical outcome prediction. Targeting PD-L1, possibly by administering miR-138-5p mimics, might be a clinically effective anti-CRC therapeutic strategy.