Merck
  • Reactivation of Human Herpes Virus-6 in the Renal Tissue of a Patient with Drug-induced Hypersensitivity Syndrome/Drug Rash with Eosinophilia and Systemic Symptoms (DIHS/DRESS).
Reactivation of Human Herpes Virus-6 in the Renal Tissue of a Patient with Drug-induced Hypersensitivity Syndrome/Drug Rash with Eosinophilia and Systemic Symptoms (DIHS/DRESS).
Internal medicine (Tokyo, Japan) (2016-07-05)
Hideharu Hagiya, Masaya Iwamuro, Takehiro Tanaka, Kou Hasegawa, Yoshihisa Hanayama, Maya Kimura, Fumio Otsuka
ABSTRACT
A 74-year-old man who had been administered trimethoprim-sulfamethoxazole for three weeks suffered from drug-induced hypersensitivity syndrome/drug rash with eosinophilia and systemic symptoms (DIHS/DRESS). In the early stage of the clinical course, he developed renal dysfunction. A renal biopsy showed granulomatous tubulointerstitial nephritis accompanying the proliferation of human herpes virus (HHV)-6 in tubular epithelial cells. With corticosteroid therapy, the systemic rash and renal function gradually improved. The present patient is the second case of DIHS/DRESS demonstrating a possible reactivation of HHV-6 in the renal tissue. The clinical role of viral reactivation in DIHS/DRESS must be further elucidated.